missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Splicing factor U2AF 26 kDa subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Splicing factor U2AF 26 kDa subunit, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7888-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001035515.1 | |
Splicing factor U2AF 26 kDa subunit Protein | |
Research Use Only | |
U2AF1L4 | |
Recombinant Protein | |
E. coli | |
MAEYLASIFG TEKDKVNCSF YFKIGVCRHG DRCSRLHNKP TFSQEVFTEL QEKYGEIEEM NVCDNLGDHL VGNVYVKFRR EEDGERAVAE LSNRWFNGQA VHGNVPEVAS ATSCICGPFP RTSRGSSMGG DPGAGHPRGS ILATIPERGT IGVPLITGMA ASEALAPLPF TPNRDRCSWQ DLSSKPPSLS CPILPRLPGS IM | |
Greater than 80% as determined by SDS-PAGE. |
199746 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |