missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Spike glycoProtein

A DNA sequence encoding the Homo sapiens (Human) Spike glycoprotein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7178-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16057580

  • 726.00€ / 200 microgramos


Fecha estimada de envío
23-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

Spike glycoProtein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
S
Recombinant Protein
E. coli
VIGDLKCTSD NINDKDTGPP PISTDTVDVT NGLGTYYVLD RVYLNTTLFL NGYYPTSGST YRNMALKGSV LLSRLWFKPP FLSDFINGIF AKVKNTKVIK DRVMYSEFPA ITIGSTFVNT SYSVVVQPRT INSTQDGDNK LQGLLEVSVC QYNMCEYPQT ICHPNLGNHR KELWHLDTGV VSCLYKRNFT YDVNADYLYF HFYQEGGTFY AYFTDTGVVT KFLFNVYLGM ALSHYYVMPL TCNSKLTLEY WVTPLTSRQY LLAFNQDGII FNAEDCMSDF MSEIKCKTQS IAPPTGVYEL NGYTVQPIAD VYRRKPNLPN CNIEAWLNDK
Videos
Documentos