missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRPB Protein
A cDNA sequence encoding the SNRPB was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13543-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SNRPB Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MTVGKSSKML QHIDYRMRCI LQDGRIFIGT FKAFDKHMNL ILCDCDEFRK IKPKNSKQAE REEKRVLGLV LLRGENLVSM TVEGPPPKDT GIARVPLAGA AGGPGIGRAA GRGIPAGVPM PQAPAGLAGP VRGVGGPSQQ VMTPQGRGTV AAAAAAATAS IAGAPTQYPP GRGGPPPPMG RGAPPPGMMG PPPGMRPPMG PPMGIPPGRG TPMGMPPPGM RPPPPGMRGP PPPGMRPPRP | |
Greater than 80% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SNRPB is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |