missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SMCP Protein

A DNA sequence encoding the Homo sapiens (Human) SMCP, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6700-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16085220

  • 675.00€ / 200 microgramos


Fecha estimada de envío
01-07-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_109588.2
SMCP Protein
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
4184
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SMCP
Recombinant Protein
E. coli
MCDQTKHSKC CPAKGNQCCP PQQNQCCQSK GNQCCPPKQN QCCQPKGSQC CPPKHNHCCQ PKPPCCIQAR CCGLETKPEV SPLNMESEPN SPQTQDKGCQ TQQQPHSPQN ESRPSK
Videos
Documentos