missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SMCP Protein
A DNA sequence encoding the Homo sapiens (Human) SMCP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6700-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_109588.2 | |
SMCP Protein | |
Research Use Only | |
SMCP | |
Recombinant Protein | |
E. coli | |
MCDQTKHSKC CPAKGNQCCP PQQNQCCQSK GNQCCPPKQN QCCQPKGSQC CPPKHNHCCQ PKPPCCIQAR CCGLETKPEV SPLNMESEPN SPQTQDKGCQ TQQQPHSPQN ESRPSK | |
Greater than 90% as determined by SDS-PAGE. |
4184 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |