missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Small nuclear ribonucleoprotein Sm D2 Protein
A DNA sequence encoding the Homo sapiens (Human) Small nuclear ribonucleoprotein Sm D2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8419-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004588.1 | |
Small nuclear ribonucleoprotein Sm D2 Protein | |
Research Use Only | |
SNRPD2 | |
Recombinant Protein | |
E. coli | |
MSLLNKPKSE MTPEELQKRE EEEFNTGPLS VLTQSVKNNT QVLINCRNNK KLLGRVKAFD RHCNMVLENV KEMWTEVPKS GKGKKKSKPV NKDRYISKMF LRGDSVIVVL RNPLIAGK | |
Greater than 90% as determined by SDS-PAGE. |
6633 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |