missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SLC4A4 Protein
A cDNA sequence encoding the SLC4A4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13522-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SLC4A4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMSTE NVEGKPSNLG ERGRARSSTF LRVVQPMFNH SIFTSAVSPA AERIRFILGEEDDSPAPPQL FTELDELLAV DGQEMEWKET ARWIKFEEKV EQGGERWSKP HVATLSLHSL FELRTCMEKG SIMLDREASS LPQLVEMIVDHQIETGLLKP ELKDKVTYTL LRKHRHQTKK SNLRSLADIG KTVSSASRMF TNPDNGSPAM THRNLTSSSL NDISDKPEKD QLKNKFMKKLPRDAEASNVL VGEVDFLDTP FIAFVRLQQA VMLGALTEVP VPTRFLFILL GPKGKAKSYH EIGRAIATLM SDEVFHDIAY KAKDRHDLIAGIDEFLDEVI VLPPGEWDPA IRIEPPKSLP SSDKRKNMYS GGENVQMNGD TPHDGGHGGG GHGDCEELQR TGRFCGGLIK DIKRKAPFFASDFYDALNIQ | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SLC4A4 protein solution contains 20mM Tris-HCl, pH 7.5, 0.5mM DTT, 0.1mM PMSF and 10% glycerol. |