missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SIRPG / SIRP gamma / CD172g Protein

A DNA sequence encoding the Homo sapiens (Human) SIRPG / SIRP gamma / CD172g, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6687-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055110

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
27-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001034597.1
SIRPG / SIRP gamma / CD172g Protein
500 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
55423
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SIRPG
Recombinant Protein
E. coli
EEELQMIQPE KLLLVTVGKT ATLHCTVTSL LPVGPVLWFR GVGPGRELIY NQKEGHFPRV TTVSDLTKRN NMDFSIRISS ITPADVGTYY CVKFRKGSPE NVEFKSGPGT EMALGAKPSA PVVLGPAART TPEHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP TGQSVAYSIR STARVVLDPW DVRSQVICEV AHVTLQGDPL RGTANLSEAI RVPPTLEVTQ QPMRVGNQVN VTCQVRKFYP QSLQLTWSEN GNVCQRETAS TLTENKDGTY NWTSWFLVNI SDQRDDVVLT CQVKHDGQLA VSKRLALEVT VHQKDQSSDA TP
Videos
Documentos