missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SIRP alpha / CD172a Protein

A DNA sequence encoding the Homo sapiens (Human) SIRP alpha / CD172a, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6684-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055080

  • 160.00€ / 10 microgramos


Fecha estimada de envío
24-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001035111.1
SIRP alpha / CD172a Protein
10 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
140885
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SIRPA
Recombinant Protein
E. coli
EEELQVIQPD KSVLVAAGET ATLRCTATSL IPVGPIQWFR GAGPGRELIY NQKEGHFPRV TTVSDLTKRN NMDFSIRIGN ITPADAGTYY CVKFRKGSPD DVEFKSGAGT ELSVRAKPSA PVVSGPAARA TPQHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP VGESVSYSIH STAKVVLTRE DVHSQVICEV AHVTLQGDPL RGTANLSETI RVPPTLEVTQ QPVRAENQVN VTCQVRKFYP QRLQLTWLEN GNVSRTETAS TVTENKDGTY NWMSWLLVNV SAHRDDVKLT CQVEHDGQPA VSKSHDLKVS AHPKEQGSNT AAENTGSNER NIY
Videos
Documentos