missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SIKE1 Protein
A cDNA sequence encoding the SIKE1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13502-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
80143 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSCTIEK ILTDAKTLLE RLREHDAAAE SLVDQSAALH RRVAAMREAG TALPDQYQED ASDMKDMSKY KPHILLSQEN TQIRDLQQEN RELWISLEEH QDALELIMSK YRKQMLQLMV AKKAVDAEPV LKAHQSHSAE IESQIDRICE MGEVMRKAVQ VDDDQFCKIQ EKLAQLELEN KELRELLSIS SESLQARKEN SMDTASQAIK | |
Greater than 85% as determined by SDS-PAGE. |
SIKE1 Protein | |
Research Use Only | |
SIKE1 | |
Human | |
His | |
SIKE1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |