missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Signal-regulatory protein beta-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Signal-regulatory protein beta-1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6685-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055090

  • 352.00€ / 50 microgramos


Fecha estimada de envío
19-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001077379.1
Signal-regulatory protein beta-1 Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
10326
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SIRPB1
Recombinant Protein
E. coli
EDELQVIQPE KSVSVAAGES ATLRCAMTSL IPVGPIMWFR GAGAGRELIY NQKEGHFPRV TTVSELTKRN NLDFSISISN ITPADAGTYY CVKFRKGSPD DVEFKSGAGT ELSVRAKPSA PVVSGPAVRA TPEHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP AGDSVSYSIH STARVVLTRG DVHSQVICEI AHITLQGDPL RGTANLSEAI RVPPTLEVTQ QPMRAENQAN VTCQVSNFYP RGLQLTWLEN GNVSRTETAS TLIENKDGTY NWMSWLLVNT CAHRDDVVLT CQVEHDGQQA VSKSYALEIS AHQKEHGSDI THEAALAPTA PL
Videos
Documentos