missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SHBG Protein
A cDNA sequence encoding the SHBG was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13492-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SHBG Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
AAQPARRARR TKLLLLLLLL LRHTRQGWAL RPVLPTQSAH DPPAVHLSNG PGQEPIAVMT FDLTKITKTS SSFEVRTWDP EGVIFYGDTN PKDDWFMLGL RDGRPEIQLH NHWAQLTVGA GPRLDDGRWH QVEVKMEGDS VLLEVDGEEV LRLRQVSGPL TSKRHPIMRI ALGGLLFPAS NLRLPLVPAL DGCLRRDSWL DKQAEISASA PTSLRSCDVE SNPGIFLPPG TQAEFNLRDI PQPHAEPWAF SLDLGLKQAA GSGHLLALGT PENPSWLSLH LQDQKVVLSS GSGPGLDLPL VLGLPLQLKL SMSRVVLSQG SKMKALALPP LGLAPLLNLW AKPQGRLFLG ALPGEDSSTS FCLNGLWAQG QRLDVDQALN RSHEIWTHSC PQSPGNGTDA SHSRGGPEQKLISEEDLNSA VDHHHHHH |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SHBG filtered solution at a concentration of 55.6microg/ml in certified FBS (Fetal Bovine Serum 5%). |