missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sesquipedalian-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Sesquipedalian-2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7760-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16050641

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001002034.2
Sesquipedalian-2 Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
150368
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FAM109B
Recombinant Protein
E. coli
MKLNERSVAH YALSDSPADH MGFLRTWGGP GTPPTPSGTG RRCWFVLKGN LLFSFESREG RAPLSLVVLE GCTVELAEAP VPEEFAFAIC FDAPGVRPHL LAAEGPAAQE AWVKVLSRAS FGYMRLVVRE LESQLQDARQ SLALQRRSSW KSVASRCKPQ APNHRAAGLE NGHCLSKDSS PVGLVEEAGS RSAGWGLAEW ELQGPASLLL GKGQSPVSPE TSCFSTLHDW YGQEIVELRQ CWQKRAQGSH SKCEEQDRP
Videos
Documentos