missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SerpinB9 / CAP-3 Protein
A DNA sequence encoding the Homo sapiens (Human) SerpinB9 / CAP-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6671-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004146.1 | |
SerpinB9 / CAP-3 Protein | |
Research Use Only | |
SerpinB9 | |
Recombinant Protein | |
E. coli | |
METLSNASGT FAIRLLKILC QDNPSHNVFC SPVSISSALA MVLLGAKGNT ATQMAQALSL NTEEDIHRAF QSLLTEVNKA GTQYLLRTAN RLFGEKTCQF LSTFKESCLQ FYHAELKELS FIRAAEESRK HINTWVSKKT EGKIEELLPG SSIDAETRLV LVNAIYFKGK WNEPFDETYT REMPFKINQE EQRPVQMMYQ EATFKLAHVG EVRAQLLELP YARKELSLLV LLPDDGVELS TVEKSLTFEK LTAWTKPDCM KSTEVEVLLP KFKLQEDYDM ESVLRHLGIV DAFQQGKADL SAMSAERDLC LSKFVHKSFV EVNEEGTEAA AASSCFVVAE CCMESGPRFC ADHPFLFFIR HNRANSILFC GRFSSP | |
Greater than 80% as determined by SDS-PAGE. |
5272 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |