missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SERPINB5 Protein
A cDNA sequence encoding the SERPINB5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13462-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SERPINB5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDALQLANSA FAVDLFKQLC EKEPLGNVLF SPICLSTSLS LAQVGAKGDT ANEIGQVLHF ENVKDIPFGF QTVTSDVNKL SSFYSLKLIK RLYVDKSLNL STEFISSTKR PYAKELETVD FKDKLEETKG QINNSIKDLT DGHFENILAD NSVNDQTKIL VVNAAYFVGK WMKKFPESET KECPFRLNKT DTKPVQMMNM EATFCMGNID SINCKIIELP FQNKHLSMFI LLPKDVEDES TGLEKIEKQL NSESLSQWTN PSTMANAKVK LSIPKFKVEK MIDPKACLEN LGLKHIFSED TSDFSGMSET KGVALSNVIH KVCLEITEDG GDSIEVPGAR ILQHKDELNA DHPFIYIIRH NKTRNIIFFG KFCSP | |
Greater than 97.0% as determined by(a) Analysis by HPLC. (b) Analysis by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.0. |