missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SerpinB2 / PAI-2 Protein
A DNA sequence encoding the Human SerpinB2 / PAI-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10161-ec-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001137290.1 | |
SerpinB2 / PAI-2 Protein | |
Research Use Only | |
SERPINB2 | |
The biologically active was determined by its inhibitory effect against single chain tPA induced cleavage of a chromogenic substrate in Imidazole Buffer at 37 C. Half maximal inhibition against 1.0 ug/ml of single chain tPA was at a concentration of 1.0 ug/ml. | |
Human | |
Untagged | |
0.2 um filtered 20 mM Tris-HCl, pH 8.0, 150 mM NaCl, 1 mM Cysteine, with 5 % Trehalose, lyophilized |
5055 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MEDLCVANTL FALNLFKHLA KASPTQNLFL SPWSISSTMA MVYMGSRGST EDQMAKVLQF NEVGANAVTP MTPENFTSCG FMQQIQKGSY PDAILQAQAA DKIHSSFRSL SSAINASTGN YLLESVNKLF GEKSASFREE YIRLCQKYYS SEPQAVDFLE CAEEARKKIN SWVKTQTKGK IPNLLPEGSV DGDTRMVLVN AVYFKGKWKT PFEKKLNGLY PFRVNSAQRT PVQMMYLREK LNIGYIEDLK AQILELPYAG DVSMFLLLPD EIADVSTGLE LLESEITYDK LNKWTSKDKM AEDEVEVYIP QFKLEEHYEL RSILRSMGME DAFNKGRANF SGMSERNDLF LSEVFHQAMV DVNEEGTEAA AGTGGVMTGR TGHGGPQFVA DHPFLFLIMH KITNCILFFG RFSSP | |
Greater than 95% as determined by SDS-PAGE and HPLC. |