missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SERPINA3 Protein

A cDNA sequence encoding the SERPINA3 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13456-HIS-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913629

  • 2583.00€ / 1 miligramo


Fecha estimada de envío
26-06-2019
Añadir a la cesta

Especificaciones

Especificaciones

SERPINA3 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Lyophilized from 0.02M NH4HCO3.
Greater than 99.0%.
Research Use Only
Recombinant Protein
E. coli
MERMLPLLAL GLLAAGFCPA VLCHPNSPLD EENLTQENQD RGTHVDLGLA SANVDFAFSL YKQLVLKAPD KNVIFSPLSI STALAFLSLG AHNTTLTEIL KGLKFNLTET SEAEIHQSFQ HLLRTLNQSS DELQLSMGNA MFVKEQLSLL DRFTEDAKRL YGSEAFATDF QDSAAAKKLI NDYVKNGTRG KITDLIKDLD SQTMMVLVNY IFFKAKWEMP FDPQDTHQSR FYLSKKKWVM VPMMSLHHLT IPYFRDEELS CTVVELKYTG NASALFILPD QDKMEEVEAM LLPETLKRWR DSLEFREIGE LYLPKFSISR DYNLNDILLQ LGIEEAFTSK ADLSGITGAR NLAVSQVVHK AVLDVFEEGT EASAATAVKI TLLSALVETR TIVRFNRPFL MIIVPTDTQN IFFMSKVTNP KQA
Videos