missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Semenogelin-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Semenogelin-2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9386-ye-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16058231

  • 1190.00€ / 200 microgramos


Fecha estimada de envío
15-01-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_002999.1
Semenogelin-2 Protein
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
6407
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SEMG2
Recombinant Protein
Yeast
QKGGSKGQLP SGSSQFPHGQ KGQHYFGQKD QQHTKSKGSF SIQHTYHVDI NDHDWTRKSQ QYDLNALHKA TKSKQHLGGS QQLLNYKQEG RDHDKSKGHF HMIVIHHKGG QAHHGTQNPS QDQGNSPSGK GLSSQCSNTE KRLWVHGLSK EQASASGAQK GRTQGGSQSS YVLQTEELVV NKQQRETKNS HQNKGHYQNV VDVREEHSSK LQTSLHPAHQ DRLQHGPKDI FTTQDELLVY NKNQHQTKNL SQDQEHGRKA HKISYPSSRT EERQLHHGEK SVQKDVSKGS ISIQTEEKIH GKSQNQVTIH SQDQEHGHKE NKISYQSSST EERHLNCGEK GIQKGVSKGS ISIQTEEQIH GKSQNQVRIP SQAQEYGHKE NKISYQSSST EERRLNSGEK DVQKGVSKGS ISIQTEEKIH GKSQNQVTIP SQDQEHGHKE NKMSYQSSST EERRLNYGGK STQKDVSQSS ISFQIEKLVE GKSQIQTPNP NQDQWSGQNA KGKSGQSADS KQDLLSHEQK GRYKQESSES HNIVITEHEV AQDDHLTQQY NEDRNPIST
Videos
Documentos