missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SELM / Selenoprotein 1
A DNA sequence encoding the Homo sapiens (Human) SELM / Selenoprotein M, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP1347-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_536355.1 | |
SELM / Selenoprotein 1 | |
Research Use Only | |
SELM | |
Recombinant Protein | |
E. coli | |
ATAYRPDWNR LSGLTRARVE TCGGSQLNRL KEVKAFVTQD IPFYHNLVMK HLPGADPELV LLGRRYEELE RIPLSEMTRE EINALVQELG FYRKAAPDAQ VPPEYVWAPA KPPEETSDHA DL | |
Greater than 97.7% as determined by SDS-PAGE. |
140606 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |