missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SELM / Selenoprotein 1

A DNA sequence encoding the Homo sapiens (Human) SELM / Selenoprotein M, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP1347-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913679

  • 1425.00€ / 500 microgramos


Fecha estimada de envío
28-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_536355.1
SELM / Selenoprotein 1
500 μg
Not determined.
Full Length of Mature Protein
Human
His
Tris-based buffer, 50% glycerol
140606
Greater than 97.7% as determined by SDS-PAGE.
Research Use Only
SELM
Recombinant Protein
E. coli
ATAYRPDWNR LSGLTRARVE TCGGSQLNRL KEVKAFVTQD IPFYHNLVMK HLPGADPELV LLGRRYEELE RIPLSEMTRE EINALVQELG FYRKAAPDAQ VPPEYVWAPA KPPEETSDHA DL
Videos
Documentos