missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Secreted Ly-6 / uPAR-related protein 1

A DNA sequence encoding the Homo sapiens (Human) Secreted Ly-6/uPAR-related protein 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6696-ye-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055190

  • 472.00€ / 100 microgramos


Fecha estimada de envío
16-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_065160.1
Secreted Ly-6 / uPAR-related protein 1
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
57152
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SLURP1
Recombinant Protein
Yeast
LKCYTCKEPM TSASCRTITR CKPEDTACMT TLVTVEAEYP FNQSPVVTRS CSSSCVATDP DSIGAAHLIF CCFRDLCNSE L
Videos
Documentos