missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100P Protein
A cDNA sequence encoding the S100P was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13408-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
S100P Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MTELEAAMGM IIDVFSRYSG SEGSTQTLTK GELKVLMEKE LPGFLQSGKD KDAVDKLLKD LDANGDAQVD FSEFIVFVAA ITSACHKYFE KAGLK | |
Greater than 95% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Phosphate buffer pH 7.2, 0.1M NaCl. |