missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100B Protein
A cDNA sequence encoding the S100B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13404-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
6285 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSELEKAMVA LIDVFHQYSG REGDKHKLKK SELKELINNE LSHFLEEIKE QEVVDKVMET LDNDGDGECD FQEFMAFVAM VTTACHEFFE HE | |
Greater than 90.0% as determined by SDS-PAGE. |
S100B Protein | |
Research Use Only | |
S100B | |
Human | |
His | |
S100B (1 mg/ml) is supplied in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |