missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A3 / S100E Protein

A DNA sequence encoding the Homo sapiens (Human) S100A3 / S100E, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6638-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054720

  • 160.00€ / 10 microgramos


Fecha estimada de envío
16-01-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_002951.1
S100A3 / S100E Protein
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6274
Greater than 80% as determined by SDS-PAGE.
Research Use Only
S100A3
Recombinant Protein
E. coli
ARPLEMS-QA VAAIVCTFQE YAGRCGDKYK LCQAELKELL QKELATWTPT EFRECDYNKF MSVLDTNKDC EVDFVEYVRS LACLCLYCHE YFKDCPSEPP CSQ
Videos
Documentos