missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A3 / S100E Protein
A DNA sequence encoding the Homo sapiens (Human) S100A3 / S100E, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6638-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002951.1 | |
S100A3 / S100E Protein | |
Research Use Only | |
S100A3 | |
Recombinant Protein | |
E. coli | |
ARPLEMS-QA VAAIVCTFQE YAGRCGDKYK LCQAELKELL QKELATWTPT EFRECDYNKF MSVLDTNKDC EVDFVEYVRS LACLCLYCHE YFKDCPSEPP CSQ | |
Greater than 80% as determined by SDS-PAGE. |
6274 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |