missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A16 Protein

A cDNA sequence encoding the S100A16 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13391-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923419

  • 3091.00€ / 1 miligramo


Fecha estimada de envío
14-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

S100A16 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
S100A16 (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 40% glycerol and 0.2M NaCl.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSDCYTELEK AVIVLVENFY KYVSKYSLVK NKISKSSFRE MLQKELNHML SDTGNRKAAD KLIQNLDANH DGRISFDEYW TLIGGITGPI AKLIHEQEQQ SSS
Videos
Documentos