missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ryanodine receptor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Ryanodine receptor 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6633-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054690

  • 160.00€ / 10 microgramos


Fecha estimada de envío
19-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000531.2
Ryanodine receptor 1 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
6261
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RYR1
Recombinant Protein
E. coli
MGDAEGEDEV QFLRTDDEVV LQCSATVLKE QLKLCLAAEG FGNRLCFLEP TSNAQNVPPD LAICCFVLEQ SLSVRALQEM LANTVEAGVE SSQGGGHRTL LYGHAILLRH AHSRMYLSCL TTSRSMTDKL AFDVGLQEDA TGEACWWTMH PASKQRSEGE KVRVGDDIIL VSVSSERYLH LSTASGELQV DASFMQTLWN MNPICSRCEE GFVTGGHVLR LFHGHMDECL TISPADSDDQ RRLVYYEGGA VCTHARSLWR LEPLRISWSG SHLRWGQPLR VRHVTTGQYL ALTEDQGLVV VDASKAHTKA TSFCFRISKE KLDVAPKRDV EGMGPPEIKY GESLCFVQHV ASGLWLTYAA PDPKALRLGV LKKKAMLHQE GHMDDALSLT RCQQEESQAA RMIHSTNGLY NQFIKSLDSF SGKPRGSGPP AGTALPIEGV ILSLQDLIIY FEPPSEDLQH EEKQSKLRSL RNRQSLFQEE GMLSMVLNCI DRLNVYTTAA HFAEFAGEEA AESWKEIVNL LYELLASLIR GNRS
Videos
Documentos