missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Runt-related transcription factor 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Runt-related transcription factor 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6632-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001026850.1 | |
Runt-related transcription factor 3 Protein | |
Research Use Only | |
RUNX3 | |
Recombinant Protein | |
E. coli | |
MRIPVDPSTS RRFTPPSPAF PCGGGGGKMG ENSGALSAQA AVGPGGRARP EVRSMVDVLA DHAGELVRTD SPNFLCSVLP SHWRCNKTLP VAFKVVALGD VPDGTVVTVM AGNDENYSAE LRNASAVMKN QVARFNDLRF VGRSGRGKSF TLTITVFTNP TQVATYHRAI KVTVDGPREP RRHRQKLEDQ TKPFPDRFGD LERLRMRVTP STPSPRGSLS TTSHFSSQPQ TPIQGTSELN PFSDPRQFDR SFPTLPTLTE SRFPDPRMHY PGAMSAAFPY SATPSGTSIS SLSVAGMPAT SRFHHTYLPP PYPGAPQNQS GPFQANPSPY HLYYGTSSGS YQFSMVAGSS SGGDRSPTRM LASCTSSAAS VAAGNLMNPS LGGQSDGVEA DGSHSNSPTA LSTPGRMDEA VWRPY | |
Greater than 90% as determined by SDS-PAGE. |
864 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |