missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RSG1 Protein
A cDNA sequence encoding the RSG1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13364-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RSG1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMARPPVP GSVVVPNWHE SAEGKEYLAC ILRKNRRRVF GLLERPVLLP PVSIDTASYK IFVSGKSGVG KTALVAKLAG LEVPVVHHET TGIQTTVVFW PAKLQASSRV VMFRFEFWDC GESALKKFDH MLLACMENTD AFLFLFSFTD RASFEDLPGQ LARIAGEAPG VVRMVIGSKF DQYMHTDVPE RDLTAFRQAW ELPLLRVKSV PGRRLADGRT LDGRAGLADV AHILNGLAEQ LWHQDQVAAG LLPNPPESAP E | |
Greater than 85.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RSG1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.5), 0.2M NaCl, 50% glycerol and 2mM DTT. |