missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RRM2B / P53R2 Protein
A DNA sequence encoding the Homo sapiens (Human) RRM2B / P53R2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7745-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001165948.1 | |
RRM2B / P53R2 Protein | |
Research Use Only | |
RRM2B | |
Recombinant Protein | |
E. coli | |
MGDPERPEAA GLDQDERSSS DTNESEIKSN EEPLLRKSSR RFVIFPIQYP DIWKMYKQAQ ASFWTAEEVD LSKDLPHWNK LKADEKYFIS HILAFFAASD GIVNENLVER FSQEVQVPEA RCFYGFQILI ENVHSEMYSL LIDTYIRDPK KREFLFNAIE TMPYVKKKAD WALRWIADRK STFGERVVAF AAVEGVFFSG SFAAIFWLKK RGLMPGLTFS NELISRDEGL HCDFACLMFQ YLVNKPSEER VREIIVDAVK IEQEFLTEAL PVGLIGMNCI LMKQYIEFVA DRLLVELGFS KVFQAENPFD FMENISLEGK TNFFEKRVSE YQRFAVMAET TDNVFTLDAD F | |
Greater than 80% as determined by SDS-PAGE. |
50484 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |