missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS20 Protein
A cDNA sequence encoding the RPS20 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13355-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RPS20 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAFKDTG KTPVEPEVAI HRIRITLTSR NVKSLEKVCA DLIRGAKEKN LKVKGPVRMP TKTLRITTRK TPCGEGSKTW DRFQMRIHKR LIDLHSPSEI VKQITSISIE PGVELIESTD AEPMDTEGQQ YTLRSVFESP GTCPF | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RPS20 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT and 1mM EDTA. |