missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPL18A Protein

A cDNA sequence encoding the RPL18A was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13342-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923269

  • 4118.00€ / 1 miligramo


Fecha estimada de envío
12-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

RPL18A Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RPL18A solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMKASGTL REYKVVGRCL PTPKCHTPPL YRMRIFAPNH VVAKSRFWYF VSQLKKMKKS SGEIVYCGQV FEKSPLRVKN FGIWLRYDSR SGTHNMYREY RDLTTAGAVT QCYRDMGARH RARAHSIQIM KVEEIAASKC RRPAVKQFHD SKIKFPLPHR VLRRQHKPRF TTKRPNTFF
Videos