missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RLBP1 Protein
A cDNA sequence encoding the RLBP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13321-HIS-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RLBP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEGVGT FRMVPEEEQE LRAQLEQLTT KDHGPVFGPC SQLPRHTLQK AKDELNEREE TREEAVRELQ EMVQAQAASG EELAVAVAER VQEKDSGFFL RFIRARKFNV GRAYELLRGY VNFRLQYPEL FDSLSPEAVR CTIEAGYPGV LSSRDKYGRV VMLFNIENWQ SQEITFDEIL QAYCFILEKL LENEETQING FCIIENFKGF TMQQAASLRT SDLRKMVDML QDSFPARFKA IHFIHQPWYF TTTYNVVKPF LKSKLLERVF VHGDDLSGFY QEIDENILPS DFGGTLPKYD GKAVAEQLFG PQAQAENTAF | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RLBP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |