missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RGS21 Protein

A cDNA sequence encoding the RGS21 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13311-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923169

  • 103.00€ / 5 microgramos


Fecha de envío estimada
del proveedor 11-04-2019
Añadir a la cesta

Especificación

Especificación

RGS21 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RGS21 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.1M NaCl.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMPVKCC FYRSPTAETM TWSENMDTLL ANQAGLDAFR IFLKSEFSEE NVEFWLACED FKKTKNADKI ASKAKMIYSE FIEADAPKEI NIDFGTRDLI SKNIAEPTLK CFDEAQKLIY CLMAKDSFPR FLKSEIYKKL VNSQQVPNHK KWLPFL
Videos