missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Retinol-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Retinol-binding protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6588-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004155.2 | |
Retinol-binding protein 1 | |
Research Use Only | |
RBP2 | |
Recombinant Protein | |
E. coli | |
TRDQNGTWEM ESNENFEGYM KALDIDFATR KIAVRLTQTK VIDQDGDNFK TKTTSTFRNY DVDFTVGVEF DEYTKSLDNR HVKALVTWEG DVLVCVQKGE KENRGWKQWI EGDKLYLELT CGDQVCRQVF KKK | |
Greater than 80% as determined by SDS-PAGE. |
5948 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |