missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Retinal dehydrogenase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Retinal dehydrogenase 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8968-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001193826.1 | |
Retinal dehydrogenase 2 Protein | |
Research Use Only | |
ALDH1A2 | |
Recombinant Protein | |
Yeast | |
MTSSKIEMPG EVKADPAALM ASLHLLPSPT PNLEIKYTKI FINNEWQNSE SGRVFPVYNP ATGEQVCEVQ EADKADIDKA VQAARLAFSL GSVWRRMDAS ERGRLLDKLA DLVERDRAVL ATMESLNGGK PFLQAFYVDL QGVIKTFRYY AGWADKIHGM TIPVDGDYFT FTRHEPIGVC GQIIPWNFPL LMFAWKIAPA LCCGNTVVIK PAEQTPLSAL YMGALIKEAG FPPGVINILP GYGPTAGAAI ASHIGIDKIA FTGSTEVGKL IQEAAGRSNL KRVTLELGGK SPNIIFADAD LDYAVEQAHQ GVFFNQGQCC TAGSRIFVEE SIYEEFVRRS VERAKRRVVG SPFDPTTEQG PQIDKKQYNK ILELIQSGVA EGAKLECGGK GLGRKGFFIE PTVFSNVTDD MRIAKEEIFG PVQEILRFKT MDEVIERANN SDFGLVAAVF TNDINKALTV SSAMQAGTVW INCYNALNAQ SPFGGFKMSG NGREMGEFGL REYSEVKTVT VKIPQKNS | |
Greater than 90% as determined by SDS-PAGE. |
8854 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |