missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Reelin Protein

A DNA sequence encoding the Homo sapiens (Human) Reelin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6597-ye-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054440

  • 805.00€ / 200 microgramos


Fecha estimada de envío
25-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_005036.2
Reelin Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
5649
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RELN
Recombinant Protein
Yeast
AAGYYPRFSP FFFLCTHHGE LEGDGEQGEV LISLHIAGNP TYYVPGQEYH VTISTSTFFD GLLVTGLYTS TSVQASQSIG GSSAFGFGIM SDHQFGNQFM CSVVASHVSH LPTTNLSFIW IAPPAGTGCV NFMATATHRG QVIFKDALAQ QLCEQGAPTD VTVHPHLAEI HSDSIILRDD FDSYHQLQLN PNIWVECNNC ETGEQCGAIM HGNAVTFCEP YGPRELITT
Videos
Documentos