missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras-related protein Rab-5C Protein

A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-5C, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6573-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054260

Cantidad Precio
1 675.00€ / 200 microgramos
5 629.51€ / 200 microgramos
Fecha estimada de envío
10-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_001238968.1
Ras-related protein Rab-5C Protein
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5878
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RAB5C
Recombinant Protein
E. coli
MAGRGGAARP NGPAAGNKIC QFKLVLLGES AVGKSSLVLR FVKGQFHEYQ ESTIGAAFLT QTVCLDDTTV KFEIWDTAGQ ERYHSLAPMY YRGAQAAIVV YDITNTDTFA RAKNWVKELQ RQASPNIVIA LAGNKADLAS KRAVEFQEAQ AYADDNSLLF METSAKTAMN VNEIFMAIAK KLPKNEPQNA TGAPGRNRGV DLQENNPASR SQCCSN
Videos
Documentos