missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras-related C3 botulinum toxin substrate 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Ras-related C3 botulinum toxin substrate 3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6576-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054280

  • 234.00€ / 50 microgramos


Fecha estimada de envío
31-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_005043.1
Ras-related C3 botulinum toxin substrate 3 Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
5881
Greater than 80% as determined by SDS-PAGE.
Research Use Only
RAC3
Recombinant Protein
E. coli
MQAIKCVVVG DGAVGKTCLL ISYTTNAFPG EYIPTVFDNY SANVMVDGKP VNLGLWDTAG QEDYDRLRPL SYPQTDVFLI CFSLVSPASF ENVRAKWYPE VRHHCPHTPI LLVGTKLDLR DDKDTIERLR DKKLAPITYP QGLAMAREIG SVKYLECSAL TQRGLKTVFD EAIRAVLCPP PVKKPGKKC
Videos
Documentos