missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAC2 Protein
A cDNA sequence encoding the RAC2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13250-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RAC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSPILGYWKI KGLVQPTRLL LEYLEEKYEE HLYERDEGDK WRNKKFELGL EFPNLPYYID GDVKLTQSMA IIRYIADKHN MLGGCPKERA EISMLEGAVL DIRYGVSRIA YSKDFETLKV DFLSKLPEML KMFEDRLCHK TYLNGDHVTH PDFMLYDALD VVLYMDPMCL DAFPKLVCFK KRIEAIPQID KYLKSSKYIA WPLQGWQATF GGGDHPPKSD LVPRGSHMQA IKCVVVGDGA VGKTCLLISY TTNAFPGEYI PTVFDNYSAN VMVDSKPVNL GLWDTAGQED YDRLRPLSYP QTDVFLICFS LVSPASYENV RAKWFPEVRH HCPSTPIILV GTKLDLRDDK DTIEKLKEKK LAPITYPQGL ALAKEIDSVK YLECSALTQR GLKTVFDEAI RAVLCPQPTR QQKRACSLL | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The RAC2 protein solution (1 mg/ml) contains 1x PBS pH 7.4. |