missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAB3IL1 Protein

A cDNA sequence encoding the RAB3IL1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13244-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922959

  • 6130.00€ / 1 miligramo


Fecha estimada de envío
09-09-2020
Añadir a la cesta

Especificaciones

Especificaciones

RAB3IL1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RAB3IL1 protein solution (0.25 mg/ml) containing Phosphate buffer saline (pH 7.4) and 50% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMWSGPPQ PDQGLPPPLA AVPVPWKSTD PCQGHRESPG ALVETSAGEE AQGQEGPAAA QLDVLRLRSS SMEIREKGSE FLKEELHRAQ KELKLKDEEC ERLSKVREQL EQELEELTAS LFEEAHKMVR EANMKQAASE KQLKEARGKI DMLQAEVTAL KTLVITSTPA SPNRELHPQL LSPTKAGPRK GHSRHKSTSS TLCPAVCPAA GHTLTPDREG KEVDTILFAE FQAWRESPTL DKTCPFLERV YREDVGPCLD FTMQELSVLV RAAVEDNTLT IEPVASQTLP TVKVAEVDCS STNTCALSGL TRTCRHRIRL GDSKSHYYIS PSSRARITAV CNFFTYIRYI QQGLVRQDAE PMFWEIMRLR KEMSLAKLGF FPQEA
Videos
Documentos