missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB14 Protein
A cDNA sequence encoding the RAB14 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13230-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RAB14 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMATAPYN YSYIFKYIII GDMGVGKSCL LHQFTEKKFM ADCPHTIGVE FGTRIIEVSG QKIKLQIWDT AGQERFRAVT RSYYRGAAGA LMVYDITRRS TYNHLSSWLT DARNLTNPNT VIILIGNKAD LEAQRDVTYE EAKQFAEENG LLFLEASAKT GENVEDAFLE AAKKIYQNIQ DGSLDLNAAE SGVQHKPSAP QGGRLTSEPQ PQREGCGC | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RAB14 protein 0.5 mg/ml is supplied in 20mM Tris-HCL, pH 8.0, 0.1M NaCl, 1mM DTT and 20% Glycerol. |