missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human QDPR Protein
A cDNA sequence encoding the QDPR was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13224-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
QDPR Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAAAAA GEARRVLVYG GRGALGSRCV QAFRARNWWV ASVDVVENEE ASASIIVKMT DSFTEQADQV TAEVGKLLGE EKVDAILCVA GGWAGGNAKS KSLFKNCDLM WKQSIWTSTI SSHLATKHLK EGGLLTLAGA KAALDGTPGM IGYGMAKGAV HQLCQSLAGK NSGMPPGAAA IAVLPVTLDT PMNRKSMPEA DFSSWTPLEF LVETFHDWIT GKNRPSSGSL IQVVTTEGRT ELTPAYF | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
QDPR protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 2mM DTT. |