missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Putative inactive neutral ceramidase B Protein

A DNA sequence encoding the Homo sapiens (Human) Putative inactive neutral ceramidase B, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5682-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916979

  • 218.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 09-04-2019
Añadir a la cesta

Especificación

Especificación

NP_001072984.1
Putative inactive neutral ceramidase B Protein
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
653308
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ASAH2B
Recombinant Protein
Yeast
MRQHRQFMDR THYLLTFSSS ETLLRLLLRI VDRAPKGRTF GDVLQPAKPE YRVGEVAEVI FVGANPKNSV QNQTHQTFLT VEKYEATSTS WQIVCNDASW ETRFYWHKGL LGLSNATVEW HIPDTAQPGI YRIRYFGHNR KQDILKPAVI LSFEGTSPAF EVVTI
Videos