missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pulmonary surfactant-associated protein A2 Protein
A DNA sequence encoding the Homo sapiens(Human) Pulmonary surfactant-associated protein A2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10008-iv-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001092138.1 | |
Pulmonary surfactant-associated protein A2 Protein | |
Research Use Only | |
SFTPA2 | |
Recombinant Protein | |
E. coli | |
EVKDVCVGSP GIPGTPGSHG LPGRDGRDGV KGDPGPPGPM GPPGETPCPP GNNGLPGAPG VPGERGEKGE AGERGPPGLP AHLDEELQAT LHDFRHQILQ TRGALSLQGS IMTVGEKVFS SNGQSITFDA IQEACARAGG RIAVPRNPEE NEAIASFVKK YNTYAYVGLT EGPSPGDFRY SDGTPVNYTN WYRGEPAGRG KEQCVEMYTD GQWNDRNCLY SRLTICEF | |
Greater than 89.6% as determined by SDS-PAGE. |
729238 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |