missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTHrP Protein

A cDNA sequence encoding the PTHrP was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13204-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922829

Cantidad Precio
1 107.00€ / 10 microgramos
10 99.50€ / 10 microgramos
Fecha estimada de envío
14-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

PTHrP Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
PthRp protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4.
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
AVSEHQLLHD KGKSIQDLRR RFFLHHLIAE IHTAEIRATS EVSPNSKPSP NTKNHPVRFG SDDEGRYLTQ ETNKVETYKE QPLKTP
Videos
Documentos