missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PSPN Protein

A DNA sequence encoding the Human PSPN, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10423-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933808

  • 668.00€ / 100 microgramos


Fecha estimada de envío
16-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_004149.1
PSPN Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
ALSGPCQLWS LTLSVAELGL GYASEEKVIF RYCAGSCPRG ARTQHGLALA RLQGQGRAHG GPCCRPTRYT DVAFLDDRHR WQRLPQLSAA ACGCGG
5623
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
PSPN
The ED50 as determined by a cell proliferation assay using human TT medullary thyroid cancer cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos