missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMD11 Protein
A DNA sequence encoding the Homo sapiens (Human) PSMD11, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8324-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001257411.1 | |
PSMD11 Protein | |
Research Use Only | |
PSMD11 | |
Recombinant Protein | |
E. coli | |
AAAAVVEFQR AQSLLSTDRE ASIDILHSIV KRDIQENDEE AVQVKEQSIL ELGSLLAKTG QAAELGGLLK YVRPFLNSIS KAKAARLVRS LLDLFLDMEA ATGQEVELCL ECIEWAKSEK RTFLRQALEA RLVSLYFDTK RYQEALHLGS QLLRELKKMD DKALLVEVQL LESKTYHALS NLPKARAALT SARTTANAIY CPPKLQATLD MQSGIIHAAE EKDWKTAYSY FYEAFEGYDS IDSPKAITSL KYMLLCKIML NTPEDVQALV SGKLALRYAG RQTEALKCVA QASKNRSLAD FEKALTDYRA ELRDDPIIST HLAKLYDNLL EQNLIRVIEP FSRVQIEHIS SLIKLSKADV ERKLSQMILD KKFHGILDQG EGVLIIFDEP PVDKTYEAAL ETIQNMSKVV DSLYNKAKKL T | |
Greater than 90% as determined by SDS-PAGE. |
5717 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |