missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human pS2 / pNR-2 / Trefoil Factor 1 Protein

A DNA sequence encoding the Human pS2 / pNR-2 / Trefoil Factor 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10178-ec-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15932988

  • 267.00€ / 20 microgramos


Fecha estimada de envío
20-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_003216.1
pS2 / pNR-2 / Trefoil Factor 1 Protein
20 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
EAQTETCTVA PRERQNCGFP GVTPSQCANK GCCFDDTVRG VPWCFYPNTI DVPPEEECEF
7031
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
TFF1
The ED50 as determined by a chemotaxis bioassay using human MCF-7 cells is less than 10 ug/ml, corresponding to a specific activity of > 100 IU/mg.
Human
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized
Videos
Documentos