missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein Wnt-5a Protein

A DNA sequence encoding the Homo sapiens (Human) Protein Wnt-5a, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6889-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16056570

  • 414.00€ / 100 microgramos


Fecha estimada de envío
13-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001243034.1
Protein Wnt-5a Protein
100 μg
Not determined.
Full Length
Human
His-Myc
Tris-based buffer, 50% glycerol
7474
Greater than 90% as determined by SDS-PAGE.
Research Use Only
WNT5A
Recombinant Protein
E. coli
IIGAQPLCSQ LAGLSQGQKK LCHLYQDHMQ YIGEGAKTGI KECQYQFRHR RWNCSTVDNT SVFGRVMQIG SRETAFTYAV SAAGVVNAMS RACREGELST CGCSRAARPK DLPRDWLWGG CGDNIDYGYR FAKEFVDARE RERIHAKGSY ESARILMNLH NNEAGRRTVY NLADVACKCH GVSGSCSLKT CWLQLADFRK VGDALKEKYD SAAAMRLNSR GKLVQVNSRF NSPTTQDLVY IDPSPDYCVR NESTGSLGTQ GRLCNKTSEG MDGCELMCCG RGYDQFKTVQ TERCHCKFHW CCYVKCKKCT EIVDQFVCK
Videos
Documentos